ethyl 3-(allylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate

Lieferant: Apollo Scientific

OR25324-1G
APOSOR25324-1GEA 125 EUR
APOSOR25324-1G
ethyl 3-(allylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
ethyl 3-(allylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
Formel: C₁₄H₁₆O₃S₂
Molecular Weight: 296,405 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085049
CAS-Nummer: 172516-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR