3-(Diethoxymethyl)-2-ethoxytetrahydro-2H-pyran

Lieferant: Apollo Scientific
Achtung

OR25325-1G
APOSOR25325-1GEA 102 EUR
APOSOR25325-1G
3-(Diethoxymethyl)-2-ethoxytetrahydro-2H-pyran
3-(Diethoxymethyl)-2-ethoxytetrahydro-2H-pyran
Formel: C₁₂H₂₄O₄
Molecular Weight: 232.32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085003
CAS-Nummer: 69549-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR