(4,5-Dichloro-1H-imidazol-1-yl)methyl (4-chlorophenyl)carbamate

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-({[(4-Chlorophenyl)carbamoyl]oxy}methyl)-4,5-dichloro-1H-imidazole

OR25372-250MG
APOSOR25372-250MGEA 111 EUR
APOSOR25372-250MG
(4,5-Dichloro-1H-imidazol-1-yl)methyl (4-chlorophenyl)carbamate
(4,5-Dichloro-1H-imidazol-1-yl)methyl (4-chlorophenyl)carbamate
Formel: C₁₁H₈Cl₃N₃O₂
Molecular Weight: 320,559 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00114451
CAS-Nummer: 650592-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR