5-(Thien-2-yl)tetrahydrothiophen-3-one

Lieferant: Apollo Scientific

Synonyme: 2,3-Dihydro-2,2'-bithiophen-4(5H)-one

OR25430-250MG
APOSOR25430-250MGEA 180 EUR
APOSOR25430-250MG
5-(Thien-2-yl)tetrahydrothiophen-3-one
5-(Thien-2-yl)tetrahydrothiophen-3-one
Formel: C₈H₈OS₂
Molecular Weight: 184,279 g/mol
Schmelzpunkt: 55…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052287
CAS-Nummer: 108372-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR