3-Phenoxythiophene-2-carboxaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: [(2-Formylthien-3-yl)oxy]benzene, 3-Phenoxy-2-thenaldehyde, 2-Formyl-3-phenoxythiophene

OR25439-1G
APOSOR25439-1GEA 95.7 EUR
APOSOR25439-1G
3-Phenoxythiophene-2-carboxaldehyde 95%
3-Phenoxythiophene-2-carboxaldehyde
Formel: C₁₁H₈O₂S
Molecular Weight: 204,245 g/mol
Siedepunkt: 184 °C (15 mm Hg)
Lagertemperatur: Kühlschrank
MDL: MFCD00178748
CAS-Nummer: 132706-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR