Methyl 2-[(3-amino-3-thioxopropyl)thio]acetate
Lieferant: Apollo Scientific
OR25440-1G
APOSOR25440-1GEA
248
EUR
APOSOR25440-1G
Methyl 2-[(3-amino-3-thioxopropyl)thio]acetate
Methyl 2-[(3-amino-3-thioxopropyl)thio]acetate
Formel:
C₆H₁₁NO₂S₂ Molecular Weight: 193,287 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052045 CAS-Nummer: 175202-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...