Di-2-thienyl sulphide 97%

Lieferant: Apollo Scientific

OR25442-1G OR25442-5G
APOSOR25442-1GEA 0 EUR
APOSOR25442-1G APOSOR25442-5G
Di-2-thienyl sulphide 97%
Di-2-thienyl sulphide
Formel: C₈H₆S₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00052047
CAS-Nummer: 3988-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR