3-Chloro-4-(phenylthio)tetrahydro-1H-1lambda~6~-thiophene-1,1-dione
Lieferant: Apollo Scientific
|
Achtung
|
OR25447-1G
APOSOR25447-1GEA
69.6
EUR
APOSOR25447-1G
3-Chloro-4-(phenylthio)tetrahydro-1H-1lambda~6~-thiophene-1,1-dione
3-Chloro-4-(phenylthio)tetrahydro-1H-1lambda~6~-thiophene-1,1-dione
Formel:
C₁₀H₁₁ClO₂S₂ Molecular Weight: 262,776 g/mol Schmelzpunkt: 78…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00174266 CAS-Nummer: 15507-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...