3-(Methylthio)thiophene

Lieferant: Apollo Scientific
Achtung

OR25457-1G
APOSOR25457-1GEA 23.2 EUR
APOSOR25457-1G
3-(Methylthio)thiophene
3-(Methylthio)thiophene
Formel: C₅H₆S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00068009
CAS-Nummer: 20731-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR