2,2-Dimethyl-2H-chromene-6-carbonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR25515-1G
APOSOR25515-1GEA
393
EUR
APOSOR25515-1G
2,2-Dimethyl-2H-chromene-6-carbonitrile 97%
2,2-Dimethyl-2H-chromene-6-carbonitrile
Formel:
C₁₂H₁₁NO Molecular Weight: 185.23 g/mol Schmelzpunkt: 45…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084978 CAS-Nummer: 33143-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...