2-Isopropyl-6-nitrophenol
Lieferant: Apollo Scientific
|
Achtung
|
OR25535-1G
OR25535-250MG
APOSOR25535-1GEA
400
EUR
APOSOR25535-1G
APOSOR25535-250MG
2-Isopropyl-6-nitrophenol
2-Isopropyl-6-nitrophenol
Formel:
C₉H₁₁NO₃ Siedepunkt: 131…135 °C/20mmHg Lagertemperatur: Raumtemperatur |
CAS-Nummer:
7545-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...