2-Isopropyl-6-nitrophenol

Lieferant: Apollo Scientific
Achtung

OR25535-1G OR25535-250MG
APOSOR25535-1GEA 400 EUR
APOSOR25535-1G APOSOR25535-250MG
2-Isopropyl-6-nitrophenol
2-Isopropyl-6-nitrophenol
Formel: C₉H₁₁NO₃
Siedepunkt: 131…135 °C/20mmHg
Lagertemperatur: Raumtemperatur
CAS-Nummer: 7545-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR