2-(Methylthio)-6-nitro-1,3-benzothiazole
Lieferant: Apollo Scientific
|
Achtung
|
OR25544-1G
OR25544-5G
APOSOR25544-1GEA
0
EUR
APOSOR25544-1G
APOSOR25544-5G
2-(Methylthio)-6-nitro-1,3-benzothiazole
2-(Methylthio)-6-nitro-1,3-benzothiazole
Formel:
C₈H₆N₂O₂S₂ Schmelzpunkt: 128…134 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00085141 CAS-Nummer: 3621-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...