3-Amino-2-brom-6-methylpyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-6-methylpyridin-3-amine, 3-Amino-2-bromo-6-picoline, 5-Amino-6-bromo-2-picoline
OR2557-1G
APOSOR2557-1GEA
21.9
EUR
APOSOR2557-1G
3-Amino-2-brom-6-methylpyridin
3-Amino-2-bromo-6-methylpyridine
Formel:
C₆H₇BrN₂ Molecular Weight: 187,04 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03095078 CAS-Nummer: 126325-53-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...