1-Isopropyluracil-5-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Cyano-2,4-dioxo-1-isopropyl-1,2,3,4-tetrahydropyrimidine, 2,4-Dioxo-1-(prop-2-yl)-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

OR25579-1G
APOSOR25579-1GEA 145 EUR
APOSOR25579-1G
1-Isopropyluracil-5-carbonitrile
1-Isopropyluracil-5-carbonitrile
Formel: C₈H₉N₃O₂
Molecular Weight: 179.176 g/mol
Schmelzpunkt: 265…274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00203975
CAS-Nummer: 57712-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR