(1,4-Dimethylpiperazin-2-yl)methanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,4-Dimethyl-2-(hydroxymethyl)piperazine

OR25585-1G
APOSOR25585-1GEA 174 EUR
APOSOR25585-1G
(1,4-Dimethylpiperazin-2-yl)methanol
(1,4-Dimethylpiperazin-2-yl)methanol
Formel: C₇H₁₆N₂O
Molecular Weight: 144.22 g/mol
Siedepunkt: 114…115 °C/ 15mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00174342
CAS-Nummer: 14675-44-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR