2-Brom-5-hydroxy-4-methoxybenzaldehyd 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR2559-5G
APOSOR2559-5GEA
21.9
EUR
APOSOR2559-5G
2-Brom-5-hydroxy-4-methoxybenzaldehyd 95%
2-Bromo-5-hydroxy-4-methoxybenzaldehyde
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol Schmelzpunkt: 95…102 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00195552 CAS-Nummer: 2973-59-3 EINECS: 426-540-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...