2,6-Dichlor-4-nitrotoluol
Lieferant: Apollo Scientific
|
Achtung
|
OR25607-1G
APOSOR25607-1GEA
0
EUR
APOSOR25607-1G
2,6-Dichlor-4-nitrotoluol
2,6-Dichlor-4-nitrotoluol
Formel:
C₇H₅Cl₂NO₂ Schmelzpunkt: 63…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00174267 CAS-Nummer: 7149-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...