3,6-Dichlor-4-methylpyridazin
Lieferant: Apollo Scientific
|
Achtung
|
OR25616-1G
APOSOR25616-1GEA
21.9
EUR
APOSOR25616-1G
3,6-Dichlor-4-methylpyridazin
3,6-Dichloro-4-methylpyridazine
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163,01 g/mol Siedepunkt: 149…151 °C/21mmHg Schmelzpunkt: 86…88 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006465 CAS-Nummer: 19064-64-3 EINECS: 242-794-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...