Methyl-5-brom-2-furoat 97%
Lieferant: Apollo Scientific
Synonyme:
5-Bromo-2-furoic acid methyll ester, 5-Bromo-2-furancarboxylic acid methyll ester, Methyl 5-Bromo-2-furancarboxylate, Methyl 5-Bromo-2-furoate, Methyl 5-bromofuran-2-carboxylate
OR25651-25G
APOSOR25651-25GEA
87
EUR
APOSOR25651-25G
Methyl-5-brom-2-furoat 97%
Methyl-5-brom-2-furoat
Formel:
C₆H₅BrO₃ Molecular Weight: 205,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092313 CAS-Nummer: 2527-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...