1-Boc-5-aminoindol 95%

Lieferant: Apollo Scientific
Achtung

OR2566-1G
APOSOR2566-1GEA 0 EUR
APOSOR2566-1G
1-Boc-5-aminoindol 95%
1-Boc-5-aminoindol
Formel: C₁₃H₁₆N₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD04114746
CAS-Nummer: 166104-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR