Diethyl 1-benzylpyrrolidine-2,5-dicarboxylate

Lieferant: Apollo Scientific
Gefahr

Synonyme: {[2,5-Bis(ethoxycarbonyl)pyrrolidin-1-yl]methyl}benzene, 1-Benzyl-2,5-bis(ethoxycarbonyl)pyrrolidine

OR25711-250MG
APOSOR25711-250MGEA 40.6 EUR
APOSOR25711-250MG
Diethyl 1-benzylpyrrolidine-2,5-dicarboxylate
Diethyl 1-benzylpyrrolidine-2,5-dicarboxylate
Formel: C₁₇H₂₃NO₄
Molecular Weight: 305.369 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00830964
CAS-Nummer: 17740-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR