2-(5-Bromo-1H-indol-3-yl)-1-ethanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromoindole-3-ethanol, 2-(5-Bromo-1H-indol-3-yl)ethanol, 5-Bromo-3-(2-hydroxyethyl)indole

OR2573-250MG
APOSOR2573-250MGEA 33.5 EUR
APOSOR2573-250MG
2-(5-Bromo-1H-indol-3-yl)-1-ethanol
2-(5-Bromo-1H-indol-3-yl)-1-ethanol
Formel: C₁₀H₁₀BrNO
Molecular Weight: 240,11 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00130167
CAS-Nummer: 32774-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR