2,3-Dioxoindoline-7-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR25737-1G
APOSOR25737-1GEA 26.1 EUR
APOSOR25737-1G
2,3-Dioxoindoline-7-carboxylic acid
2,3-Dioxoindoline-7-carboxylic acid
Formel: C₉H₅NO₄
Schmelzpunkt: 268…275 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00612492
CAS-Nummer: 25128-35-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR