3-(Naphth-1-yl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(1H-Pyrazol-3-yl)naphthalene

OR2574-250MG
APOSOR2574-250MGEA 92.8 EUR
APOSOR2574-250MG
3-(Naphth-1-yl)-1H-pyrazole
3-(Naphth-1-yl)-1H-pyrazole
Formel: C₁₃H₁₀N₂
Molecular Weight: 194.232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01569443
CAS-Nummer: 150433-19-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR