Boc-glycine methyl ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Boc-glycine Methyl Ester, Boc-Gly-OMe

OR25753-100G
APOSOR25753-100GEA 55.1 EUR
APOSOR25753-100G
Boc-glycine methyl ester 97%
Boc-glycine methyl ester
Formel: C₈H₁₅NO₄
Molecular Weight: 189,2 g/mol
Siedepunkt: 190 °C
Dichte: 1,28 g/cm³ (25 °C)
Flammpunkt: 109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038267
CAS-Nummer: 31954-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR