2-Chloro-4-nitro-1H-imidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-5-nitro-1H-imidazole

OR2577-1G OR2577-25G OR2577-5G
APOSOR2577-1GEA 21.9 EUR
APOSOR2577-1G APOSOR2577-25G APOSOR2577-5G
2-Chloro-4-nitro-1H-imidazole
2-Chloro-4-nitro-1H-imidazole
Formel: C₃H₂ClN₃O₂
Molecular Weight: 147.518 g/mol
Schmelzpunkt: 214…216 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03419295
CAS-Nummer: 57531-37-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR