4-Chloro-N-ethyl-6-(1-phenyl-1H-indol-3-yl)-1,3,5-triazin-2-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[4-Chloro-6-(ethylamino)-1,3,5-triazin-2-yl]-1-phenyl-1H-indole, {3-[4-Chloro-6-(ethylamino)-1,3,5-triazin-2-yl]-1H-indol-1-yl}benzene

OR25770-10MG
APOSOR25770-10MGEA 43.5 EUR
APOSOR25770-10MG
4-Chloro-N-ethyl-6-(1-phenyl-1H-indol-3-yl)-1,3,5-triazin-2-amine
4-Chloro-N-ethyl-6-(1-phenyl-1H-indol-3-yl)-1,3,5-triazin-2-amine
Formel: C₁₉H₁₆ClN₅
Molecular Weight: 349,817 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00123281
CAS-Nummer: 500587-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR