Diethyl-2-cyano-3-oxosuccinate
Lieferant: Apollo Scientific
|
Achtung
|
OR25781-500MG
APOSOR25781-500MGEA
21.9
EUR
APOSOR25781-500MG
Diethyl-2-cyano-3-oxosuccinate
Diethyl-2-cyano-3-oxosuccinate
Formel:
C₉H₁₁NO₅ Molecular Weight: 213,19 g/mol Schmelzpunkt: 88…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026853 CAS-Nummer: 134541-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...