N-Benzyl-2,3-dichloro-6-nitroaniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(2,3-Dichloro-6-nitrophenyl)benzylamine, 2-(Benzylamino)-3,4-dichloronitrobenzene
OR25862-25MG
APOSOR25862-25MGEA
21.9
EUR
APOSOR25862-25MG
N-Benzyl-2,3-dichloro-6-nitroaniline
N-Benzyl-2,3-dichloro-6-nitroaniline
Formel:
C₁₃H₁₀Cl₂N₂O₂ Molecular Weight: 297,137 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00174601 CAS-Nummer: 680212-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...