2-Hydroxy-6-methylisonicotinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-6-methylpyridine-4-carboxylic acid

OR25900-250MG
APOSOR25900-250MGEA 21.9 EUR
APOSOR25900-250MG
2-Hydroxy-6-methylisonicotinsäure
2-Hydroxy-6-methylisonicotinsäure
Formel: C₇H₇NO₃
Molecular Weight: 153,15 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682019
CAS-Nummer: 86454-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR