Ethyl-2-(benzylamino)-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific

OR25901-1G
APOSOR25901-1GEA 87 EUR
APOSOR25901-1G
Ethyl-2-(benzylamino)-1,3-thiazole-5-carboxylate
Ethyl-2-(benzylamino)-1,3-thiazole-5-carboxylate
Formel: C₁₃H₁₄N₂O₂S
Molecular Weight: 262,33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03086199
CAS-Nummer: 342394-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR