2,5-Dimethyl-3-thiophensulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
OR25904-250MG
APOSOR25904-250MGEA
75.4
EUR
APOSOR25904-250MG
2,5-Dimethyl-3-thiophensulfonylchlorid
2,5-Dimethyl-3-thiophensulfonylchlorid
Formel:
C₆H₇ClO₂S₂ Siedepunkt: 120…122 °C/3mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD03086201 CAS-Nummer: 97272-04-3 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...