6-(4-Methylpiperazin-1-yl)nicotinic acid

Lieferant: Apollo Scientific
Achtung

OR2591-1G OR2591-250MG
APOSOR2591-1GEA 301 EUR
APOSOR2591-1G APOSOR2591-250MG
6-(4-Methylpiperazin-1-yl)nicotinic acid
6-(4-Methylpiperazin-1-yl)nicotinic acid
Formel: C₁₁H₁₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07376841
CAS-Nummer: 132521-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR