Ethyl 1-(2-chloroacetyl)piperidine-4-carboxylate

Lieferant: Apollo Scientific
Gefahr

OR25929-5G
APOSOR25929-5GEA 267 EUR
APOSOR25929-5G
Ethyl 1-(2-chloroacetyl)piperidine-4-carboxylate
Formel: C₁₀H₁₆ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01480849
CAS-Nummer: 318280-71-6
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR