Ethyl-6-chlorimidazo[1,2-b]pyridazin-2-carboxylat

Lieferant: Apollo Scientific

OR25936-5G
APOSOR25936-5GEA 79.9 EUR
APOSOR25936-5G
Ethyl-6-chlorimidazo[1,2-b]pyridazin-2-carboxylat
Ethyl-6-chlorimidazo[1,2-b]pyridazin-2-carboxylat
Formel: C₉H₈ClN₃O₂
Schmelzpunkt: 155…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086230
CAS-Nummer: 64067-99-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR