3-Hydroxythiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR2594-250MG
APOSOR2594-250MGEA
171
EUR
APOSOR2594-250MG
3-Hydroxythiophene-2-carboxylic acid
3-Hydroxythiophene-2-carboxylic acid
Formel:
C₅H₄O₃S Schmelzpunkt: 106…108 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09414719 CAS-Nummer: 5118-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...