ethyl 2-(5-bromo-2-thienyl)-1,3-thiazole-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR25943-1G
APOSOR25943-1GEA
319
EUR
APOSOR25943-1G
ethyl 2-(5-bromo-2-thienyl)-1,3-thiazole-4-carboxylate
ethyl 2-(5-bromo-2-thienyl)-1,3-thiazole-4-carboxylate
Formel:
C₁₀H₈BrNO₂S₂ Molecular Weight: 318.21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407345 CAS-Nummer: 423768-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...