1,2-Benzisoxazol-3-amin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzo[d]isoxazol-3-amine, 3-Amino-1,2-benzisoxazole
OR25956-100MG
APOSOR25956-100MGEA
21.9
EUR
APOSOR25956-100MG
1,2-Benzisoxazol-3-amin 98%
1,2-Benzisoxazol-3-amin
Formel:
C₇H₆N₂O Molecular Weight: 134,1400 g/mol Schmelzpunkt: 110 - 112 °C |
MDL:
MFCD03407353 CAS-Nummer: 36216-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...