4-Methoxy-1,2-benzisoxazol-3-amin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methoxybenzo[d]isoxazol-3-amine, 3-Amino-4-methoxy-1,2-benzisoxazole

OR25957-250MG
APOSOR25957-250MGEA 21.9 EUR
APOSOR25957-250MG
4-Methoxy-1,2-benzisoxazol-3-amin
4-Methoxy-1,2-benzisoxazol-3-amin
Formel: C₈H₈N₂O₂
Molecular Weight: 164,160 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407354
CAS-Nummer: 177995-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR