2,4-Dimethoxybenzolsulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,4-Dimethoxybenzolsulfochlorid, 2,4-Dimethoxybenzolsulfonsäurechlorid, 2,4-Dimethoxybenzol-1-sulfonylchlorid
OR25958-250MG
APOSOR25958-250MGEA
21.9
EUR
APOSOR25958-250MG
2,4-Dimethoxybenzolsulfonylchlorid
2,4-Dimethoxybenzolsulfonylchlorid
Formel:
C₈H₉ClO₄S Molecular Weight: 236,68 g/mol Schmelzpunkt: 71…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060683 CAS-Nummer: 63624-28-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...