4-Methylthiazole-2-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methyl-1,3-thiazole-2-carboxylic acid

OR2596-1G
APOSOR2596-1GEA 0 EUR
APOSOR2596-1G
4-Methylthiazole-2-carboxylic acid 95%
4-Methylthiazole-2-carboxylic acid
Formel: C₅H₅NO₂S
Molecular Weight: 143,16 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD07377486
CAS-Nummer: 14542-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR