3-(4-Methoxyphenyl)-5-methyl-4-isoxazolcarbonsäure 97%

Lieferant: Apollo Scientific
Achtung

OR25961-1G OR25961-250MG OR25961-5G
APOSOR25961-1GEA 174 EUR
APOSOR25961-1G APOSOR25961-250MG APOSOR25961-5G
3-(4-Methoxyphenyl)-5-methyl-4-isoxazolcarbonsäure 97%
3-(4-Methoxyphenyl)-5-methylisoxazole-4-carboxylic acid
Formel: C₁₂H₁₁NO₄
Schmelzpunkt: 190…195 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407357
CAS-Nummer: 93041-45-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR