1-[3-Amino-4-(methylamino)phenyl]ethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Amino-4-(methylamino)phenyl)ethanone, 3'-Amino-4'-(methylamino)acetophenone

OR25963-100MG
APOSOR25963-100MGEA 21.9 EUR
APOSOR25963-100MG
1-[3-Amino-4-(methylamino)phenyl]ethanone
1-[3-Amino-4-(methylamino)phenyl]ethanone
Formel: C₉H₁₂N₂O
Schmelzpunkt: 119…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407301
CAS-Nummer: 18076-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR