2,2-Dimethylchroman-6-sulphonyl chloride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR25965-250MG
APOSOR25965-250MGEA
168
EUR
APOSOR25965-250MG
2,2-Dimethylchroman-6-sulphonyl chloride 95%
2,2-Dimethylchroman-6-sulphonyl chloride
Formel:
C₁₁H₁₃ClO₃S Schmelzpunkt: 92…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407324 CAS-Nummer: 131880-55-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...