Ethyl-1-cyclopropyl-4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate

Lieferant: Apollo Scientific

OR25975-1G
APOSOR25975-1GEA 149 EUR
APOSOR25975-1G
Ethyl-1-cyclopropyl-4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Ethyl-1-cyclopropyl-4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Formel: C₁₃H₁₇NO₃
Molecular Weight: 235,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03659714
CAS-Nummer: 423768-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR