5-Nitro-1-benzothiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR25982-1G
APOSOR25982-1GEA
0
EUR
APOSOR25982-1G
5-Nitro-1-benzothiophene-2-carboxylic acid
5-Nitro-1-benzothiophene-2-carboxylic acid
Formel:
C₉H₅NO₄S Schmelzpunkt: 238…241 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01159700 CAS-Nummer: 6345-55-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...