3-(2-Methylpyrimidin-4-yl)benzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR25988-250MG
APOSOR25988-250MGEA 290 EUR
APOSOR25988-250MG
3-(2-Methylpyrimidin-4-yl)benzenesulphonyl chloride
3-(2-Methylpyrimidin-4-yl)benzenesulphonyl chloride
Formel: C₁₁H₉ClN₂O₂S
Molecular Weight: 268.72 g/mol
Schmelzpunkt: 174…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03659718
CAS-Nummer: 465514-07-2
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR