2-(4-Benzylpiperidin-1-yl)ethanamine

Lieferant: Apollo Scientific
Gefahr

OR25989-1G
APOSOR25989-1GEA 0 EUR
APOSOR25989-1G
2-(4-Benzylpiperidin-1-yl)ethanamine
2-(4-Benzylpiperidin-1-yl)ethanamine
Formel: C₁₄H₂₂N₂
Molecular Weight: 218,34 g/mol
Siedepunkt: 130…131 °C/3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03659719
CAS-Nummer: 25842-32-4
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR