4-Brom-1,3-dimethyl-1H-pyrazol-5-carbonsäure

Lieferant: Apollo Scientific
Achtung

OR25990-1G
APOSOR25990-1GEA 46.4 EUR
APOSOR25990-1G
4-Brom-1,3-dimethyl-1H-pyrazol-5-carbonsäure
4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid
Formel: C₆H₇BrN₂O₂
Molecular Weight: 219,0360 g/mol
Schmelzpunkt: 230…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02090873
CAS-Nummer: 5775-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR